1,2-Bis(diphenylphosphino)ethane nickel(II) chloride ≥98%
Supplier: BeanTown Chemical
Synonyms:
Dichloro[bis(1,2-diphenylphosphino)ethane]nickel(II)
219950-1G
219950-5G
BT219950-1GEA
21
USD
BT219950-1G
BT219950-5G
1,2-Bis(diphenylphosphino)ethane nickel(II) chloride ≥98%
1,2-Bis(diphenylphosphino)ethane nickel(II) chloride
CAS: 14647-23-5; MDL No: MFCD00013313
Powder; Linear Formula: NiCl2[(C6H5)2PCH2CH2P(C6H5)2]; Molecular Formula: C26H24Cl2NiP2; MW: 528.02
Melting Point: 263-265°
- Powder
Formula:
C₂₆H₂₄Cl₂NiP₂ MW: 528.016 g/mol Melting Pt: 263…265 °C |
MDL Number:
MFCD00013313 CAS Number: 14647-23-5 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...