Bis(aminoguanidinium) sulfate ≥98%
Supplier: BeanTown Chemical
219930-100G
219930-25G
BT219930-100GEA
100.03
USD
BT219930-100G
BT219930-25G
Bis(aminoguanidinium) sulfate ≥98%
Bis(aminoguanidinium)sulphate
CAS: 996-19-0; EC No: 213-628-7; MDL No: MFCD00035665
Solid; Linear Formula: NH2C(NHNH2)=NH·0.5H2SO4; Molecular Formula: CH6N4·0.5H2O4S; MW: 123.13
Melting Point: 200°
Density (g/mL): 1.36
- Solid
Formula:
C₂H₁₄N₈O₄S Melting Pt: 200 °C Density: 1.36 g/mL |
MDL Number:
MFCD00035665 CAS Number: 996-19-0 EINECS: 213-628-7 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...