N-Boc-glycine (boc-gly-OH) ≥98%

Supplier: BeanTown Chemical

219900-100G 219900-25G 219900-5G
BT219900-100GEA 304 USD
BT219900-100G BT219900-25G BT219900-5G
N-Boc-glycine (boc-gly-OH) ≥98%
N-Boc-glycine (Boc-Gly-OH)

CAS: 4530-20-5; MDL No: MFCD00002690 Powder; Linear Formula: (CH3)3COCONHCH2COOH; Molecular Formula: C7H13NO4; MW: 175.18 Melting Point: 86-89°


  • Powder

Formula: C₇H₁₃NO₄
Melting Pt: 86…89 °C
MDL Number: MFCD00002690
CAS Number: 4530-20-5

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR