2-Phenylpropionic acid ≥98%

Supplier: BeanTown Chemical
Warning

Synonyms: (±)-2-Phenylpropionic acid, (±)-Hydratropic acid, DL-2-Phenylpropionic acid, (±)-2-Phenylpropanoic acid

219885-100G 219885-25G 219885-5G
BT219885-100GEA 373 USD
BT219885-100G BT219885-25G BT219885-5G
2-Phenylpropionic acid ≥98%
2-Phenylpropionic acid

CAS: 492-37-5; EC No: 207-752-0; MDL No: MFCD00002650 Liquid; Molecular Formula: C9H10O2; MW: 150.17 Boiling Point: 260-262°; Flash point: 110°C (230°F) Density (g/mL): 1.1; Refractive Index: 1.522

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 260…262 °C
Density: 1.1
Flash Pt: 110 °C
MDL Number: MFCD00002650
CAS Number: 492-37-5
EINECS: 207-752-0

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR