Titanium(IV) oxide anatase ≥99.5%
Supplier: BeanTown Chemical
Synonyms:
Pigment White 6, Titanium dioxide anatase, Anatase
219880-100G
219880-25G
BT219880-100GEA
268
USD
BT219880-100G
BT219880-25G
Titanium(IV) oxide anatase ≥99.5%
Titanium(IV) oxide anatase
CAS: 1317-70-0; EC No: 215-280-1; MDL No: MFCD00011269; RTECS: BV6550000
3-6mm Sintered Pieces; Linear Formula: TiO2; MW: 79.90
Melting Point: 1825°
Density (g/mL): 3.9
- RTECS: BV6550000
- 3-6mm Sintered Pieces
Formula:
TiO₂ MW: 79.88 g/mol Melting Pt: 1825 °C Density: 3.9 g/mL |
MDL Number:
MFCD00011269 CAS Number: 1317-70-0 EINECS: 215-280-1 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...