2-Aminobenzimidazole ≥97%

Supplier: BeanTown Chemical

219875-100G 219875-25G 219875-5G
BT219875-100GEA 275 USD
BT219875-100G BT219875-25G BT219875-5G
2-Aminobenzimidazole ≥97%
2-Aminobenzimidazole

CAS: 934-32-7; EC No: 213-280-6; MDL No: MFCD00005596; RTECS: DD5775000 Solid; Molecular Formula: C7H7N3; MW: 133.15 Melting Point: 226-230°


  • RTECS: DD5775000
  • Solid

Formula: C₇H₇N₃
Melting Pt: 226…230 °C
MDL Number: MFCD00005596
CAS Number: 934-32-7
EINECS: 213-280-6

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR