2,4,6-Trihydroxybenzoic acid monohydrate ≥94%
Supplier: BeanTown Chemical
219830-100G
219830-25G
219830-500G
BT219830-100GEA
147
USD
BT219830-100G
BT219830-25G
BT219830-500G
2,4,6-Trihydroxybenzoic acid monohydrate ≥94%
2,4,6-Trihydroxybenzoic acid monohydrate
CAS: 71989-93-0; EC No: 201-467-5; MDL No: MFCD00149095; RTECS: DH8910000
Powder; Molecular Formula: C7H6O5·H2O; MW: 188.13
Melting Point: 205° (decomposes)
- RTECS: DH8910000
- Powder
Formula:
C₇H₆O₅·1H₂O Melting Pt: 205 °C (dec.) |
MDL Number:
MFCD00149095 CAS Number: 71989-93-0 EINECS: 201-467-5 |
Specification Test Results
Assay | 94% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...