Bismuth triacetate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Acetic acid bismuth (III) salt, Bismuth (III) acetate, Bismuth(III) acetate
219805-250G
219805-50G
BT219805-250GEA
338
USD
BT219805-250G
BT219805-50G
Bismuth triacetate ≥99%
Bismuth triacetate
CAS: 22306-37-2; EC No: 244-904-5; MDL No: MFCD00015629
Crystalline; Linear Formula: Bi(OOCCH3)3; Molecular Formula: C6H9BiO6; MW: 386.11
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
(H₃CCOO)₃Bi MW: 386.11 g/mol |
MDL Number:
MFCD00015629 CAS Number: 22306-37-2 EINECS: 244-904-5 |
Specification Test Results
Assay | 99% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...