(S)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid ≥99%

Supplier: BeanTown Chemical

Synonyms: (S)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid, (2S)-(-)-2-Methoxy-2-phenyl-3,3,3-trifluoropropanoic acid

219800-2G 219800-500MG
BT219800-2GEA 324 USD
BT219800-2G BT219800-500MG
(S)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid ≥99%
(S)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid

CAS: 17257-71-5; EC No: 241-292-1; MDL No: MFCD00064200 Solid; Molecular Formula: C10H9F3O3 ; MW: 234.18 Melting Point: 46-49°; Boiling Point: 95-97°/0.05 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.303; Optical Rotation: [α]18/D -72°, c = 1.6 in methanol Hygroscopic


  • Optical rotation: [α]18/D -72°, c = 1.6 in methanol
  • Hygroscopic
  • Solid

Formula: C₁₀H₉F₃O₃
MW: 234.17 g/mol
Boiling Pt: 95…97 °C/0.05 mmHg
Melting Pt: 46…49 °C
Density: 1.303 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00064200
CAS Number: 17257-71-5
EINECS: 241-292-1

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR