2-Amino-6-bromopyridine ≥98%

Supplier: BeanTown Chemical

Synonyms: 6-Bromo-2-pyridylamine, 6-Bromopyridin-2-amine

219795-25G 219795-5G
BT219795-25GEA 253 USD
BT219795-25G BT219795-5G
2-Amino-6-bromopyridine ≥98%
2-Amino-6-bromopyridine

CAS: 19798-81-3; MDL No: MFCD00137843 Powder; Molecular Formula: C5H5BrN2 ; MW: 173.01 Melting Point: 88-91°


  • Powder

Formula: C₅H₅BrN₂
MW: 173.01 g/mol
Melting Pt: 88…91 °C
MDL Number: MFCD00137843
CAS Number: 19798-81-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR