Dichloro(1,5-cyclooctadiene)palladium(II)
Supplier: BeanTown Chemical
Synonyms:
1,5-Cyclooctadienepalladium(II) Dichloride
219785-1G
219785-5G
BT219785-1GEA
110
USD
BT219785-1G
BT219785-5G
Dichloro(1,5-cyclooctadiene)palladium(II)
Dichloro(1,5-cyclooctadiene)palladium(II)
CAS: 12107-56-1; EC No: 235-161-8; MDL No: MFCD00012412
Crystalline; Molecular Formula: C8H12Cl2Pd; MW: 285.51
Melting Point: 210° (decomposes)
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
C₈H₁₂Cl₂Pd MW: 285.50 g/mol Melting Pt: 210 °C (dec.) |
MDL Number:
MFCD00012412 CAS Number: 12107-56-1 EINECS: 235-161-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...