Ethyl pyrrole-2-carboxylate ≥98%

Supplier: BeanTown Chemical

Synonyms: Ethyl 1H-pyrrole-2-carboxylate

219765-25G 219765-5G
BT219765-25GEA 359 USD
BT219765-25G BT219765-5G
Ethyl pyrrole-2-carboxylate ≥98%
Ethyl pyrrole-2-carboxylate

CAS: 2199-43-1; MDL No: MFCD00817049 Solid; Molecular Formula: C7H9NO2; MW: 139.16 Melting Point: 31-39°; Boiling Point: 70°/0.8 mmHg


  • Solid

Formula: C₇H₉NO₂
MW: 139.16 g/mol
Boiling Pt: 70 °C/0.8 mmHg
Melting Pt: 31…39 °C
MDL Number: MFCD00817049
CAS Number: 2199-43-1

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR