5-Methylisatin ≥97%
Supplier: BeanTown Chemical
Synonyms:
5-Methylindoline-2,3-dione, 5-Methyl-1H-indole-2,3-dione
219755-1G
219755-25G
219755-5G
BT219755-1GEA
37
USD
BT219755-1G
BT219755-25G
BT219755-5G
5-Methylisatin ≥97%
5-Methylisatin
CAS: 608-05-9; EC No: 210-152-1; MDL No: MFCD00005721; RTECS: NL7939300
Solid; Molecular Formula: C9H7NO2; MW: 161.16
Melting Point: 180° (decomposes)
- RTECS: NL7939300
- Solid
Formula:
C₉H₇NO₂ MW: 161.16 g/mol Melting Pt: 180 °C (dec.) |
MDL Number:
MFCD00005721 CAS Number: 608-05-9 EINECS: 210-152-1 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...