Diphenylmethylene-glycine benzyl ester ≥95%

Supplier: BeanTown Chemical

Synonyms: N-(Diphenylmethylene) glycine benzyl ester

219705-1G 219705-5G
BT219705-1GEA 43 USD
BT219705-1G BT219705-5G
Diphenylmethylene-glycine benzyl ester ≥95%
Diphenylmethylene-glycine benzyl ester

CAS: 81477-91-0 Solid; Molecular Formula: C22H19NO2; MW: 329.39


  • Solid

Formula: C₂₂H₁₉NO₂
MW: 329.3900 g/mol
MDL Number: MFCD08062206
CAS Number: 81477-91-0

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR