Europium(III) oxide ≥99.99%

Supplier: BeanTown Chemical

219685-25G 219685-5G
BT219685-25GEA 715 USD
BT219685-25G BT219685-5G
Europium(III) oxide ≥99.99%
Europium(III) oxide

CAS: 1308-96-9; EC No: 215-165-6; MDL No: MFCD00010997 45-60nm APS Powder, S. A. 14-18 m2/g; Molecular Formula: Eu2O3; MW: 351.92 Density (g/mL): 7.42 Hygroscopic


  • Hygroscopic
  • 45-60nm APS Powder, S. A. 14-18 m²/g

Formula: Eu₂O₃
MW: 351.92 g/mol
Melting Pt: 2325
Density: 7.42 g/mL
MDL Number: MFCD00010997
CAS Number: 1308-96-9
EINECS: 215-165-6
Merck Index: 14,03904

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR