Diethyl ethoxymethylenemalonate ≥98%

Supplier: BeanTown Chemical

Synonyms: Diethyl 2-(ethoxymethylene)malonate

219660-250G 219660-50G
BT219660-250GEA 46 USD
BT219660-250G BT219660-50G
Diethyl ethoxymethylenemalonate ≥98%
Diethyl ethoxymethylenemalonate

CAS: 87-13-8; EC No: 201-725-7; MDL No: MFCD00009148; RTECS: OO1100000 Liquid; Molecular Formula: C10H16O5; MW: 216.23 Boiling Point: 279-281°; Flash point: 144°C (291°F) Density (g/mL): 1.07; Refractive Index: 1.462


  • RTECS: OO1100000
  • Refractive index: 1.462
  • Liquid

Formula: C₁₀H₁₆O₅
MW: 216.23 g/mol
Boiling Pt: 279…281 °C
Density: 1.07 g/mL
Flash Pt: 144 °C (291 °F)
MDL Number: MFCD00009148
CAS Number: 87-13-8
EINECS: 201-725-7

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR