3,5-Di-tert-butyl-2-hydroxybenzaldehyde ≥99%

Supplier: BeanTown Chemical

Synonyms: 3,5-Di-tert-butylsalicylaldehyde

219600-100G 219600-25G 219600-5G
BT219600-100GEA 732 USD
BT219600-100G BT219600-25G BT219600-5G
3,5-Di-tert-butyl-2-hydroxybenzaldehyde ≥99%
3,5-Di-tert-butyl-2-hydroxybenzaldehyde

CAS: 37942-07-7; EC No: 000-000-0; MDL No: MFCD00191998 Crystalline; Molecular Formula: C15H22O2; MW: 234.33 Melting Point: 59-61° Air Sensitive


  • Air Sensitive
  • Crystalline

Formula: C₁₅H₂₂O₂
MW: 234.34 g/mol
Melting Pt: 59…61 °C
MDL Number: MFCD00191998
CAS Number: 37942-07-7

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR