Z-Pyr-OH ≥98%
Supplier: BeanTown Chemical
219590-25G
219590-5G
BT219590-25GEA
260
USD
BT219590-25G
BT219590-5G
Z-Pyr-OH ≥98%
Z-Pyr-OH
CAS: 32159-21-0; EC No: 250-936-0; MDL No: MFCD00037352
Solid; Molecular Formula: C13H13NO5; MW: 263.25
Melting Point: 128-130°
- Solid
Formula:
C₁₃H₁₃NO₅ Melting Pt: 128…130 °C |
MDL Number:
MFCD00037352 CAS Number: 32159-21-0 EINECS: 250-936-0 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...