Sodium butyrate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Butyric acid sodium salt, Sodium Butyrate
219565-100G
219565-500G
BT219565-100GEA
40.51
USD
BT219565-100G
BT219565-500G
Sodium butyrate ≥98%
Sodium butyrate
CAS: 156-54-7; EC No: 205-857-6; MDL No: MFCD00002816; RTECS: ET6400000
Powder; Linear Formula: CH3(CH2)2COONa; Molecular Formula: C4H7NaO2; MW: 110.09
Melting Point: 250-253°
Hygroscopic
- RTECS: ET6400000
- Hygroscopic
- Powder
Formula:
C₄H₇NaO₂ MW: 110.09 g/mol Melting Pt: 250…253 °C |
MDL Number:
MFCD00002816 CAS Number: 156-54-7 EINECS: 205-857-6 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...