2-N-Propyl-4-methyl-6-(1'-methylbenzimidazole-2-yl)benzimidazole ≥95%

Supplier: BeanTown Chemical

219535-1G 219535-5G
BT219535-1GEA 31 USD
BT219535-1G BT219535-5G
2-N-Propyl-4-methyl-6-(1'-methylbenzimidazole-2-yl)benzimidazole ≥95%
2-N-Propyl-4-methyl-6-(1'-methylbenzimidazole-2-yl)benzimidazole

CAS: 152628-02-9; MDL No: MFCD03840857 Powder; Molecular Formula: C19H20N4; MW: 304.39


  • Powder

Formula: C₁₉H₂₀N₄
MDL Number: MFCD09029193
CAS Number: 152628-02-9

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR