Ammonium hexachloropalladate(IV) ≥99.9% (trace metals basis)
Supplier: BeanTown Chemical
219530-1G
219530-5G
BT219530-1GEA
50
USD
BT219530-1G
BT219530-5G
Ammonium hexachloropalladate(IV) ≥99.9% (trace metals basis)
Ammonium hexachloropalladate(IV)
CAS: 19168-23-1; EC No: 242-854-9; MDL No: MFCD00015959; RTECS: BP5390000
Crystalline; Linear Formula: (NH4)2PdCl6; MW: 355.21
Hygroscopic
- RTECS: BP5390000
- Hygroscopic
- Crystalline
Formula:
H₈Cl₆N₂Pd |
MDL Number:
MFCD00015959 CAS Number: 19168-23-1 EINECS: 242-854-9 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...