Germanium ≥99.999% (trace metals basis), powder -100 mesh

Supplier: BeanTown Chemical

219500-10G 219500-50G
BT219500-10GEA 146 USD
BT219500-10G BT219500-50G
Germanium ≥99.999% (trace metals basis), powder -100 mesh
Germanium

CAS: 7440-56-4; EC No: 231-164-3; MDL No: MFCD00085310; RTECS: LY5200000 Powder; Molecular Formula: Ge; MW: 72.64 Melting Point: 937°; Boiling Point: 2830° Density (g/mL): 5.35


  • RTECS: LY5200000
  • Powder

Formula: Ge
MW: 72.61 g/mol
Boiling Pt: 2830 °C
Melting Pt: 937 °C
Density: 5.35 g/mL
MDL Number: MFCD00085310
CAS Number: 7440-56-4
EINECS: 231-164-3
UN: 3089
ADR: 4.1,III

Order Now

Specification Test Results

Assay 99.999%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR