3-(Hydroxymethyl)benzyl alcohol ≥98%
Supplier: BeanTown Chemical
219495-1G
219495-25G
219495-5G
BT219495-1GEA
24
USD
BT219495-1G
BT219495-25G
BT219495-5G
3-(Hydroxymethyl)benzyl alcohol ≥98%
3-(Hydroxymethyl)benzyl alcohol
CAS: 626-18-6; EC No: 210-934-2; MDL No: MFCD00004649
Solid; Molecular Formula: C8H10O2 ; MW: 138.16
Melting Point: 56-60°; Boiling Point: 154-159°/13 mmHg
- Solid
Formula:
C₈H₁₀O₂ Boiling Pt: 154…159 °C/13 mmHg Melting Pt: 56…60 °C |
MDL Number:
MFCD00004649 CAS Number: 626-18-6 EINECS: 210-934-2 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...