Tungsten trioxide ≥99.995%
Supplier: BeanTown Chemical
Synonyms:
Tungsten (VI) oxide
219475-100G
219475-25G
BT219475-100GEA
359
USD
BT219475-100G
BT219475-25G
Tungsten trioxide ≥99.995%
Tungsten trioxide
CAS: 1314-35-8; EC No: 215-231-4; MDL No: MFCD00011466; RTECS: YO7760000
Powder; Linear Formula: WO3; MW: 231.85
Melting Point: 1472°; Boiling Point: 1837°
Density (g/mL): 7.16
Moisture Sensitive
- RTECS: YO7760000
- Moisture Sensitive
- Powder
Formula:
O₃W MW: 231.84 Boiling Pt: 1837 °C Melting Pt: 1473 Density: 7.16 g/mL |
MDL Number:
MFCD00011466 CAS Number: 1314-35-8 EINECS: 215-231-4 Merck Index: 12,09947 |
Specification Test Results
Assay | 99.995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...