(4-Tolyloxy)acetic acid ≥98%

Supplier: BeanTown Chemical

219415-25G 219415-5G
BT219415-25GEA 53 USD
BT219415-25G BT219415-5G
(4-Tolyloxy)acetic acid ≥98%
(4-Tolyloxy)acetic acid

CAS: 940-64-7; EC No: 213-374-7; MDL No: MFCD00014365 Powder; Linear Formula: CH3C6H4OCH2COOH; Molecular Formula: C9H10O3; MW: 166.18 Melting Point: 137-142°


  • Powder

Formula: (C₆H₄(CH₃))OCH₂COOH
Melting Pt: 137…142 °C
MDL Number: MFCD00014365
CAS Number: 940-64-7
EINECS: 213-374-7

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR