(4-Chloro-2,6-difluorophenyl)acetic acid ≥97%
Supplier: BeanTown Chemical
219350-1G
219350-250MG
BT219350-1GEA
268
USD
BT219350-1G
BT219350-250MG
(4-Chloro-2,6-difluorophenyl)acetic acid ≥97%
(4-Chloro-2,6-difluorophenyl)acetic acid
CAS: 537033-55-9; MDL No: MFCD03094467
Powder; Molecular Formula: C8H5ClF2O2; MW: 206.58
Melting Point: 135-139°
- Powder
Formula:
C₈H₅ClF₂O₂ Melting Pt: 135…139 °C |
MDL Number:
MFCD03094467 CAS Number: 537033-55-9 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...