2,6-Difluoro-m-toluic acid ≥97%

Supplier: BeanTown Chemical

Synonyms: 2,6-Difluoro-3-methylbenzoic acid

219335-1G 219335-5G
BT219335-1GEA 54 USD
BT219335-1G BT219335-5G
2,6-Difluoro-m-toluic acid ≥97%
2,6-Difluoro-m-toluic acid

CAS: 32890-88-3; MDL No: MFCD01631332 Powder; Molecular Formula: C8H6F2O2 ; MW: 172.13


  • Powder

Formula: C₈H₆F₂O₂
MW: 172.13 g/mol
MDL Number: MFCD01631332
CAS Number: 32890-88-3

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR