2-Fluoro-5-iodopyridine ≥97%
Supplier: BeanTown Chemical
219280-1G
219280-5G
BT219280-1GEA
39
USD
BT219280-1G
BT219280-5G
2-Fluoro-5-iodopyridine ≥97%
2-Fluoro-5-iodopyridine
CAS: 171197-80-1; MDL No: MFCD03095301
Solid; Molecular Formula: C5H3FIN ; MW: 222.99
Melting Point: 33-37°; Flash point: 95°C (203°F)
Light Sensitive
- Light Sensitive
- Solid
Formula:
C₅H₃FIN Melting Pt: 33…37 °C Flash Pt: 95 °C (203 °F) |
MDL Number:
MFCD03095301 CAS Number: 171197-80-1 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...