6-Chlorooxindole ≥98%

Supplier: BeanTown Chemical

Synonyms: 6-Chloro-2-oxindole

219275-100G 219275-25G 219275-5G
BT219275-100GEA 548 USD
BT219275-100G BT219275-25G BT219275-5G
6-Chlorooxindole ≥98%
6-Chlorooxindole

CAS: 56341-37-8; MDL No: MFCD00209962 Crystalline/Powder; Molecular Formula: C8H6ClNO ; MW: 167.59 Melting Point: 195-199°


  • Crystalline/Powder

Formula: C₈H₆ClNO
MW: 167.59 g/mol
Melting Pt: 195…199 °C
MDL Number: MFCD00209962
CAS Number: 56341-37-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR