5-Methoxy-1-methyl-1H-indole-3-carbaldehyde ≥98%
Supplier: BeanTown Chemical
Synonyms:
5-Methoxy-1-methylindole-3-carboxaldehyde
219270-1G
219270-250MG
BT219270-1GEA
170
USD
BT219270-1G
BT219270-250MG
5-Methoxy-1-methyl-1H-indole-3-carbaldehyde ≥98%
5-Methoxy-1-methyl-1H-indole-3-carbaldehyde
CAS: 39974-94-2; MDL No: MFCD01578324
Solid; Molecular Formula: C11H11NO2; MW: 189.22
Melting Point: 133-136°
Air Sensitive
- Air Sensitive
- Solid
Formula:
C₁₁H₁₁NO₂ MW: 189.214 g/mol Melting Pt: 133…136 °C |
MDL Number:
MFCD01578324 CAS Number: 39974-94-2 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...