3-Fluorophenylboronic acid ≥97%
Supplier: BeanTown Chemical
Synonyms:
3-Fluorobenzeneboronic acid, (3-Fluorophenyl)boronic acid, Dihydroxy(3-fluorophenyl)borane, (3-Fluorophenyl)metaboric acid
219230-1G
219230-25G
219230-5G
BT219230-1GEA
30
USD
BT219230-1G
BT219230-25G
BT219230-5G
3-Fluorophenylboronic acid ≥97%
3-Fluorophenylboronic acid
CAS: 768-35-4; MDL No: MFCD00236042
Crystalline/Powder; Molecular Formula: C6H6BFO2; MW: 139.92
Melting Point: 214-218°
- Crystalline/Powder
Formula:
C₆H₆BFO₂ MW: 139.92 g/mol Melting Pt: 214…218 °C |
MDL Number:
MFCD00236042 CAS Number: 768-35-4 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...