Capric acid ≥99%

Supplier: BeanTown Chemical

Synonyms: Decylic acid, Decanoic acid

219205-1KG 219205-250G 219205-5KG
BT219205-1KGEA 148.47 USD
BT219205-1KG BT219205-250G BT219205-5KG
Capric acid ≥99%
Capric acid

CAS: 334-48-5; EC No: 206-376-4; MDL No: MFCD00004441; RTECS: HD9100000 Crystalline; Linear Formula: CH3(CH2)8CO2H; Molecular Formula: C10H20O2; MW: 172.27 Melting Point: 27-32°; Boiling Point: 268-270°; Flash point: 110°C (230°F) Density (g/mL): 0.893


  • RTECS: HD9100000
  • Crystalline

Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
Boiling Pt: 268…270 °C
Melting Pt: 27…32 °C
Density: 0.893 g/mL
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00004441
CAS Number: 334-48-5
EINECS: 206-376-4

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR