Potassium methanesulfonate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Potassiummethanesulphonate, Methanesulphonic acid potassium salt
219185-100G
219185-25G
219185-500G
BT219185-100GEA
130
USD
BT219185-100G
BT219185-25G
BT219185-500G
Potassium methanesulfonate ≥99%
Potassium methanesulphonate
CAS: 2386-56-3; EC No: 219-202-7; MDL No: MFCD00070544; RTECS: PB2675000
Powder/Lump; Linear Formula: CH3SO3K; MW: 134.19
Hygroscopic
- RTECS: PB2675000
- Hygroscopic
- Powder/Lump
Formula:
CH₃KO₃S MW: 134.2 g/mol |
MDL Number:
MFCD00070544 CAS Number: 2386-56-3 EINECS: 219-202-7 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...