Sodium L(+)-ascorbate ≥99%

Supplier: BeanTown Chemical

Synonyms: Sodium ascorbate, L(+)-Ascorbic acid sodium salt, L-Ascorbic acid sodium salt, Sodium (R)-2-((S)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Acidum ascorbicum sodium salt

219180-100G 219180-500G
BT219180-100GEA 34.2 USD
BT219180-100G BT219180-500G
Sodium L(+)-ascorbate ≥99%
Sodium L(+)-ascorbate

CAS: 134-03-2; EC No: 205-126-1; MDL No: MFCD00082340; RTECS: CI7671000 Solid; Molecular Formula: C6H7NaO6; MW: 198.11 Melting Point: 220° (decomposes)


  • RTECS: CI7671000
  • Solid

Formula: C₆H₇NaO₆
MW: 198.11 g/mol
Melting Pt: 220 °C (dec.)
MDL Number: MFCD00082340
CAS Number: 134-03-2
EINECS: 205-126-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d142b281353c28f2a72502b60fece03e926023d&vid=988032859afe97984b0fc1b8bfd43cff093f099a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR