3,4-Difluoro-4'-(4-ethylcyclohexyl)biphenyl ≥97%

Supplier: BeanTown Chemical

Synonyms: 3,4-Difluoro-4'-(trans-4-ethylcyclohexyl)biphenyl

219165-1G 219165-5G
BT219165-1GEA 75 USD
BT219165-1G BT219165-5G
3,4-Difluoro-4'-(4-ethylcyclohexyl)biphenyl ≥97%
3,4-Difluoro-4'-(4-ethylcyclohexyl)biphenyl

CAS: 134412-18-3; MDL No: MFCD12828121 Powder; Molecular Formula: C20H22F2; MW: 300.39 Melting Point: 68-69°


  • Powder

Formula: C₂₀H₂₂F₂
MW: 300.393 g/mol
Melting Pt: 68…69 °C
MDL Number: MFCD12828121
CAS Number: 134412-18-3

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR