Sodium 4-bromobenzenesulfinate dihydrate ≥97%

Supplier: BeanTown Chemical

219160-1G 219160-5G
BT219160-1GEA 26 USD
BT219160-1G BT219160-5G
Sodium 4-bromobenzenesulfinate dihydrate ≥97%
Sodium 4-bromobenzenesulphinate dihydrate

CAS: 175278-64-5; MDL No: MFCD00210139 Powder; Molecular Formula: C6H4BrNaO2S·2H2O; MW: 279.09


  • Powder

Formula: C₆H₄BrNaO₂S·2H₂O
MDL Number: MFCD00210139
CAS Number: 175278-64-5

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR