2,6-Dibromotoluene ≥98%

Supplier: BeanTown Chemical

219155-1G 219155-25G 219155-5G
BT219155-1GEA 43 USD
BT219155-1G BT219155-25G BT219155-5G
2,6-Dibromotoluene ≥98%
2,6-Dibromotoluene

CAS: 69321-60-4; EC No: 000-000-0; MDL No: MFCD00013524 Liquid; Molecular Formula: C7H6Br2; MW: 249.94 Density (g/mL): 1.812


  • Liquid

Formula: C₇H₆Br₂
Density: 1.812 g/mL
MDL Number: MFCD00013524
CAS Number: 69321-60-4

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR