ε-Caprolactam ≥99%

Supplier: BeanTown Chemical

Synonyms: Azepan-2-one, 2-Oxohexamethyleneimine, 6-Aminocaproic acid lactam

219135-100G 219135-2.5KG 219135-500G
BT219135-100GEA 37.18 USD
BT219135-100G BT219135-2.5KG BT219135-500G
ε-Caprolactam ≥99%
ε-Caprolactam

CAS: 105-60-2; EC No: 203-313-2; MDL No: MFCD00006936; RTECS: CM3675000 Crystalline; Molecular Formula: C6H11NO; MW: 113.16 Melting Point: 68-71°; Boiling Point: 270°; Flash point: 152°C (305°F) Density (g/mL): 1.02 Hygroscopic


  • RTECS: CM3675000
  • Hygroscopic
  • Crystalline

Formula: C₆H₁₁NO
MW: 113.16 g/mol
Boiling Pt: 270 °C
Melting Pt: 68…71 °C
Density: 1.02 g/mL
Flash Pt: 152 °C (305 °F)
MDL Number: MFCD00006936
CAS Number: 105-60-2
EINECS: 203-313-2

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR