Sodium (R)-(+)-α-[(3-methoxy-1-methyl-3-oxo-1-propenyl)amino]-1,4-cyclohexadiene-1-acetate ≥98%
Supplier: BeanTown Chemical
219100-1G
219100-5G
BT219100-1GEA
197
USD
BT219100-1G
BT219100-5G
Sodium (R)-(+)-α-[(3-methoxy-1-methyl-3-oxo-1-propenyl)amino]-1,4-cyclohexadiene-1-acetate ≥98%
Sodium (R)-(+)-α-[(3-methoxy-1-methyl-3-oxo-1-propenyl)amino]-1,4-cyclohexadiene-1-acetate
CAS: 26774-89-0; MDL No: MFCD00196811
Powder; Molecular Formula: C13H16NNaO4; MW: 273.26
Melting Point: 240° (decomposes)
Optical Rotation: [α]20/D +83°, c = 1 in H2O
- Optical rotation: [α]20/D +83°, c = 1 in H2O
- Powder
Formula:
C₁₃H₁₆NNaO₄ MW: 273.264 g/mol Melting Pt: 240 °C (dec.) |
CAS Number:
26774-89-0 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...