2,2-Dimethyl-1,3-dioxan-5-one ≥90%, Technical Grade
Supplier: BeanTown Chemical
219090-1G
219090-250MG
BT219090-1GEA
182.13
USD
BT219090-1G
BT219090-250MG
2,2-Dimethyl-1,3-dioxan-5-one ≥90%, Technical Grade
2,2-Dimethyl-1,3-dioxan-5-one
CAS: 74181-34-3; MDL No: MFCD00671514
Liquid; Molecular Formula: C6H10O3; MW: 130.15
Air Sensitive, Moisture Sensitive
- Air Sensitive, Moisture Sensitive
- Liquid
Formula:
C₆H₁₀O₃ |
MDL Number:
MFCD00671514 CAS Number: 74181-34-3 |
Specification Test Results
Assay | 90% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...