3-Carboxyphenylboronic acid

Supplier: BeanTown Chemical

Synonyms: 3-Boronobenzoic acid, 3-Carboxybenzeneboronic acid

219055-1G 219055-5G
BT219055-1GEA 58 USD
BT219055-1G BT219055-5G
3-Carboxyphenylboronic acid
3-Carboxyphenylboronic acid

CAS: 25487-66-5; MDL No: MFCD00036833; RTECS: CY8750000 Crystalline/Powder; Molecular Formula: C7H7BO4 ; MW: 165.94 Melting Point: 243-247°


  • RTECS: CY8750000
  • Crystalline/Powder

Formula: C₇H₇BO₄
MW: 165.94 g/mol
Melting Pt: 243…247 °C
MDL Number: MFCD00036833
CAS Number: 25487-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR