2-Bromo-5-chloroanisole ≥98%
Supplier: BeanTown Chemical
Synonyms:
1-Bromo-4-chloro-2-methoxybenzene
219040-25G
219040-5G
BT219040-25GEA
523
USD
BT219040-25G
BT219040-5G
2-Bromo-5-chloroanisole ≥98%
2-Bromo-5-chloroanisole
CAS: 174913-09-8; MDL No: MFCD03790889
Liquid; Molecular Formula: C7H6BrClO; MW: 221.48
- Liquid
Formula:
C₇H₆BrClO MW: 221.48 g/mol |
MDL Number:
MFCD03790889 CAS Number: 174913-09-8 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...