2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid) ≥98%

Supplier: BeanTown Chemical

219005-100G 219005-25G 219005-500G
BT219005-100GEA 115.4 USD
BT219005-100G BT219005-25G BT219005-500G
2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid) ≥98%
2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid)

CAS: 2033-24-1; EC No: 217-992-8; MDL No: MFCD00006638; RTECS: JH0800000 Crystalline; Molecular Formula: C6H8O4; MW: 144.13 Melting Point: 92-96°


  • RTECS: JH0800000
  • Crystalline

Formula: C₆H₈O₄
Melting Pt: 92…96 °C
MDL Number: MFCD00006638
CAS Number: 2033-24-1
EINECS: 217-992-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR