Dichloro(ethylenediamine)palladium(II)
Supplier: BeanTown Chemical
Synonyms:
Dichloro(1,2-diaminoethane)palladium
218950-1G
218950-5G
BT218950-1GEA
113.98
USD
BT218950-1G
BT218950-5G
Dichloro(ethylenediamine)palladium(II)
Dichloro(ethylenediamine)palladium(II)
CAS: 15020-99-2; MDL No: MFCD00044991
Crystalline; Linear Formula: PdCl2(H2NCH2CH2NH2); Molecular Formula: C2H8Cl2N2Pd; MW: 237.41
Melting Point: 278°
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
PdCl₂(H₂NCH₂CH₂NH₂) MW: 237.41 g/mol Melting Pt: 278 °C |
MDL Number:
MFCD00044991 CAS Number: 15020-99-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...