Ethyl-2-methylacetoacetate ≥97%

Supplier: BeanTown Chemical

218910-100G 218910-25G 218910-500G
BT218910-100GEA 116 USD
BT218910-100G BT218910-25G BT218910-500G
Ethyl-2-methylacetoacetate ≥97%
Ethyl-2-methylacetoacetate

CAS: 609-14-3; EC No: 210-179-9; MDL No: MFCD00009164 Liquid; Molecular Formula: C7H12O3 ; MW: 144.17 Boiling Point: 187°; Flash point: 63°C (145°F) Density (g/mL): 1.019; Refractive Index: 1.418


  • Refractive index: 1.418
  • Liquid

Formula: C₇H₁₂O₃
Boiling Pt: 187 °C
Density: 1.019 g/mL
Flash Pt: 63 °C (145 °F)
MDL Number: MFCD00009164
CAS Number: 609-14-3
EINECS: 210-179-9

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR