Calcium sulfate, anhydrous ≥99.99% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Calcium sulphate (anhydrous) anhydrite, Gypsum

218895-1G 218895-5G
BT218895-1GEA 23 USD
BT218895-1G BT218895-5G
Calcium sulfate, anhydrous ≥99.99% (trace metals basis)
Calcium sulphate

CAS: 7778-18-9; EC No: 231-900-3; MDL No: MFCD00010912; RTECS: WS6920000 Powder; Linear Formula: CaSO4; MW: 136.14 Hygroscopic


  • RTECS: WS6920000
  • Hygroscopic
  • Powder

Formula: CaO₄S
MW: 136.14 g/mol
MDL Number: MFCD00010912
CAS Number: 7778-18-9
EINECS: 231-900-3

Order Now

Specification Test Results

Assay 99.99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR