Bis(2-butoxyethyl)ether ≥99%

Supplier: BeanTown Chemical

Synonyms: Diethylene glycol dibutyl ether, 2,2-Dibutoxyethyl ether

218860-100ML 218860-25ML 218860-500ML
BT218860-100MLEA 40 USD
BT218860-100ML BT218860-25ML BT218860-500ML
Bis(2-butoxyethyl)ether ≥99%
Bis(2-butoxyethyl)ether

CAS: 112-73-2; EC No: 204-001-9; MDL No: MFCD00009459; RTECS: KN0350000 Liquid; Linear Formula: [CH3(CH2)3OCH2CH2]2O; Molecular Formula: C12H26O3; MW: 218.33 Melting Point: -60°; Boiling Point: 256°; Flash point: 118°C (244°F) Density (g/mL): 0.885; Refractive Index: 1.423


  • RTECS: KN0350000
  • Refractive index: 1.423
  • Liquid

Formula: C₁₂H₂₆O₃
MW: 218.34 g/mol
Boiling Pt: 256 °C
Melting Pt: –60 °C
Density: 0.885 g/mL
Flash Pt: 118 °C (244 °F)
MDL Number: MFCD00009459
CAS Number: 112-73-2
EINECS: 204-001-9

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR