5-Bromo-2-fluoro-3-pyridineboronic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 5-Bromo-2-fluoro-3-pyridylboronic, 5-Bromo-2-fluoro-3-pyridinylboronic acid, 5-Bromo-2-fluoropyridine-3-boronic acid

218850-1G 218850-250MG 218850-5G
BT218850-1GEA 251 USD
BT218850-1G BT218850-250MG BT218850-5G
5-Bromo-2-fluoro-3-pyridineboronic acid ≥98%
5-Bromo-2-fluoro-3-pyridineboronic acid

CAS: 501435-91-2; MDL No: MFCD04039317 Solid; Molecular Formula: C5H4BBrFNO2; MW: 219.81 Melting Point: 300°


  • Solid

Formula: C₅H₄BBrFNO₂
Melting Pt: >300 °C
MDL Number: MFCD04039317
CAS Number: 501435-91-2

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR