Water for spectrophotometry
Supplier: BeanTown Chemical
Synonyms:
Hydrogen oxide
218845-1L
218845-4L
218845-4X1L
BT218845-1LEA
37
USD
BT218845-1L
BT218845-4L
BT218845-4X1L
Water for spectrophotometry
Water
CAS: 7732-18-5; EC No: 231-791-2; MDL No: MFCD00011332
Liquid; Molecular Formula: H2O ; MW: 18.02
Melting Point: 0°; Boiling Point: 100°
Density (g/mL): 1; Refractive Index: 1.333
- Refractive index: 1.333
- Liquid
Formula:
H₂O MW: 18.02 g/mol Boiling Pt: 100 Melting Pt: 0 Density: 1 g/mL |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 EINECS: 231-791-2 Merck Index: 13,10098 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...