Theophylline-7-acetic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-(1,3-Dimethyl-2,6-dioxo-1,2,3,6-tetrahydro-7H-purin-7-yl)acetic acid

218835-100G 218835-25G 218835-500G
BT218835-100GEA 83.98 USD
BT218835-100G BT218835-25G BT218835-500G
Theophylline-7-acetic acid ≥98%
Theophylline-7-acetic acid

CAS: 652-37-9; EC No: 211-490-2; MDL No: MFCD00022832; RTECS: UO7451300 Powder; Molecular Formula: C9H10N4O4; MW: 238.20 Melting Point: 270-272°


  • RTECS: UO7451300
  • Powder

Formula: C₉H₁₀N₄O₄
MW: 238.20 g/mol
Melting Pt: 270…272 °C
MDL Number: MFCD00022832
CAS Number: 652-37-9
EINECS: 211-490-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR